Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) sulfide, 99.999% (metals basis)

Catalog No. AA4546318
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4546318 50 g
AA4546330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4546318 Supplier Thermo Scientific Chemicals Supplier No. 04546318
Add to Cart

CAS: 1345-07-9 | Bi2S3 | 514.14 g/mol

Bismuth(III) sulfide is mainly used in solar cells, photodiode array and infrared spectroscopy. It is also applied in the manufacture of other bismuth compounds, additives for free-cutting steels and micro-electronics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1345-07-9
Molecular Formula Bi2S3
Molecular Weight (g/mol) 514.14
MDL Number MFCD00014204
InChI Key YNRGZHRFBQOYPP-UHFFFAOYSA-N
IUPAC Name dibismuth(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Bi+3].[Bi+3]

Specifications

Color Gray
Physical Form Lump
Quantity 50 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 514.15
Density 7.7 g/cm3
Chemical Name or Material Bismuth(III) sulfide
Hazard Category H315-H319-H335-H303: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
EINECSNumber 215-716-0
RTECSNumber EB3105000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.