Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Indium Lead Tin eutectic ingot, alloy 136, 99.9% (metals basis)

Catalog No. AA1814322
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1814322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1814322 Supplier Thermo Scientific Chemicals Supplier No. 01814322
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bismuth Indium Lead Tin eutectic ingot
Name Note Alloy 136
Melting Point 58°C
Avg. Mol. Wt. or Mol. Wt. Range Bi:In:Pb:Sn; 49:21:18:12 wt%
Quantity 100 g
Odor Odorless
Form Ingot
Assay Percent Notes (metals basis)
MDL Number MFCD00210606
Percent Purity 99.9%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.