Learn More
Hach Company Bismuth Nitrate Digital Titrator Cartridge, 0.0200 M
Supplier:  Hach Company 2434501
Encompass
For determination of Total Chelant by Digital Titrator titration. Digits x 0.188 = mg/L Total Chelant (as Na4EDTA) in 50 mL sample.
Catalog No. 50-199-5984