Learn More
Electron Microscopy Sciences Bismuth Stain Kit 2
SDP
Catalog No. 50980205
Encompass_Preferred
Bismuth specifically stains Golgi beads and interchromatin granules after fixation with glutaraldehyde, whereas it stains nucleoli, basic protein, and biogenic amines after formaldehyde fixation. Bismuth staining of sections is relatively nonspecific: it can stain glycogen, lysosomes, ribosomes, polysaccharides, ferritin, and DNA. M.A. Hayat, "Basic Techniques for TEM" p. 184 (1986). Enhancing Contrast in General (Riva, 1974). Kit contains 5x10 ml Solution A(Sodium tartrate in NaOH) 5x200mg Ingredient B (Bismuth subnitrate).
Catalog No. 50-980-205 Supplier Electron Microscopy Sciences Supplier No. 11434
Bismuth specifically stains Golgi beads and interchromatin granules after fixation with glutaraldehyde, whereas it stains nucleoli, basic protein, and biogenic amines after formaldehyde fixation. Bismuth staining of sections is relatively nonspecific: it can stain glycogen, lysosomes, ribosomes, polysaccharides, ferritin, and DNA. M.A. Hayat, "Basic Techniques for TEM" p. 184 (1986). Enhancing Contrast in General (Riva, 1974). Kit contains 5x10 ml Solution A(Sodium tartrate in NaOH) 5x200mg Ingredient B (Bismuth subnitrate).