Learn More
Electron Microscopy Sciences Bismuth Stain Kit 3
SDP
Catalog No. 50980206
Encompass_Preferred
Bismuth specifically stains Golgi beads and interchromatin granules after fixation with glutaraldehyde, whereas it stains nucleoli, basic protein, and biogenic amines after formaldehyde fixation. Bismuth staining of sections is relatively nonspecific: it can stain glycogen, lysosomes, ribosomes, polysaccharides, ferritin, and DNA. M.A. Hayat, "Basic Techniques for TEM" p. 184 (1986). Staining of Mucosubstances & Polysaccharides (Ainsworth et al., 1972). Kit contains: 10x10 ml Solution A (Periodic acid, Ethanol, Sodium acetate) 10x10 ml Solution B (Sodium tartrate, NaOH, Bismuth subnitrate).
Catalog No. 50-980-206 Supplier Electron Microscopy Sciences Supplier No. 11436
Bismuth specifically stains Golgi beads and interchromatin granules after fixation with glutaraldehyde, whereas it stains nucleoli, basic protein, and biogenic amines after formaldehyde fixation. Bismuth staining of sections is relatively nonspecific: it can stain glycogen, lysosomes, ribosomes, polysaccharides, ferritin, and DNA. M.A. Hayat, "Basic Techniques for TEM" p. 184 (1986). Staining of Mucosubstances & Polysaccharides (Ainsworth et al., 1972). Kit contains: 10x10 ml Solution A (Periodic acid, Ethanol, Sodium acetate) 10x10 ml Solution B (Sodium tartrate, NaOH, Bismuth subnitrate).