Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Standard, 10,000μg/mL in water w/Dilute Nitric Acid, Ultra Scientific

Catalog No. USICP183
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USICP183 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USICP183 Supplier Agilent Technologies Supplier No. ICP183
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7440-69-9 , 7697-37-2 , 7732-18-5
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]

Specifications

Physical Form Liquid
Quantity 125 mL
Concentration or Composition (by Analyte or Components) 10, 000μg/mL
Chemical Name or Material Bismuth Standard, 10, 000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.