Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Standard, 1mL = 1mg Bi, 1000ppm Bi, Ricca Chemical
SDP

Catalog No. 100016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
100016 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 100016 Supplier Ricca Chemical Company Supplier No. 100016

Chemical Identifiers

CAS 7440-69-9 , 7697-37-2 , 7732-18-5
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]

Specifications

Packaging Poly natural
Concentration or Composition Notes 1000ppm Bi
Equivalence 1 mL = 1 mg Bi
Physical Form Liquid
Color Colorless
Quantity 500 mL
Chemical Name or Material Bismuth Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.