Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Bismuth Subcarbonate USP
SDP

Catalog No. 14185550 Shop All Medisca Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-550 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-185-550 Supplier Medisca Supplier No. 065908
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bismuth Subcarbonate USP is a white or almost white powder. Alternative name(s): Bismuth carbonate oxide

Bismuth Subcarbonate USP is a white or almost white powder.

Alternative name(s): Bismuth carbonate oxide

Specifications

Chemical Name or Material Bismuth Subcarbonate USP
Packaging Jar, 1500mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 5892-10-4
Type Drug Standard
Molecular Weight (g/mol) 509.97
Quantity 500 g
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.