Learn More
Electron Microscopy Sciences Bismuth Subnitrate, Crystalline 25 GR
SDP
Catalog No. 50980207
Encompass_Preferred
(Bismuth Nitrate, Basic; Bismuth Oxynitrate Hydrate)
CAS #1304-85-4 4BiNO₃(OH₂).BiO(OH) F.W. 1461.99
Bismuth staining for light and electron microscopy. M .Locke and P. Huie, Tissue & Cell, 9, 347 (1977); G.L. Brown & M. Locke, ibd. 10, 365 (1978)
Catalog No. 50-980-207 Supplier Electron Microscopy Sciences Supplier No. 11450
(Bismuth Nitrate, Basic; Bismuth Oxynitrate Hydrate)
CAS #1304-85-4 4BiNO₃(OH₂).BiO(OH) F.W. 1461.99
Bismuth staining for light and electron microscopy. M .Locke and P. Huie, Tissue & Cell, 9, 347 (1977); G.L. Brown & M. Locke, ibd. 10, 365 (1978)