Learn More
Apexbio Technology LLC Bismuth Subsalicylate 14882-18-9 5g
SDP
Catalog No. 5000096210
Encompass_Preferred
Bismuth subsalicylate (CAS 14882-18-9) is a small molecule compound clinically employed in the management of gastrointestinal disturbances including diarrhea heartburn indigestion and nausea Its pharmacological activity involves a dual mechanism the bismuth component exerts antimicrobial and protective effects on the gastrointestinal mucosa while the salicylate moiety inhibits intestinal prostaglandin synthesis thereby reducing inflammation and intestinal secretions Bismuth subsalicylate is utilized in biomedical research to investigate gastrointestinal barrier function host microbe interactions and inflammatory pathways relevant to digestive disorders
Catalog No. 50-000-96210 Supplier Apexbio Technology LLC Supplier No. A83825000
Bismuth subsalicylate (CAS 14882-18-9) is a small molecule compound clinically employed in the management of gastrointestinal disturbances including diarrhea heartburn indigestion and nausea Its pharmacological activity involves a dual mechanism the bismuth component exerts antimicrobial and protective effects on the gastrointestinal mucosa while the salicylate moiety inhibits intestinal prostaglandin synthesis thereby reducing inflammation and intestinal secretions Bismuth subsalicylate is utilized in biomedical research to investigate gastrointestinal barrier function host microbe interactions and inflammatory pathways relevant to digestive disorders