Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Subsalicylate, Purified, 56.5-60.5%, Spectrum™ Chemical
SDP

Catalog No. 18609122 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-122 50 kg Amber Glass Bottle
18-609-118 125 g Amber Glass Bottle
18-609-119 500 g Amber Glass Bottle
18-609-120 2.5 kg Amber Glass Bottle
18-609-121 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-609-122 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. BI15250KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14882-18-9

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Percent Purity 56.5 to 60.5%
Grade Purified
Loss on Drying 0.01
Packaging Amber Glass Bottle
Chemical Name or Material Bismuth Subsalicylate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.