Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Subsalicylate, USP, Spectrum™ Chemical
SDP

Catalog No. 18601019 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-019 25 g Poly Bottle
18-601-020 125 g Poly Bottle
18-601-021 500 g Poly Bottle
18-601-022 2.5 kg Poly Bottle
18-601-023 12 kg Poly Bottle
18-601-024 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-601-019 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B170325GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

B1703, 14882-18-9, C7H5BiO4

Spectrum™ Chemical Bismuth Subsalicylate, USP is an antacid medication used to treat temporary discomforts of the stomach and gastrointestinal tract. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 14882-18-9
Molecular Formula C7H6BiO4
Molecular Weight (g/mol) 363.10
InChI Key QBWLKDFBINPHFT-UHFFFAOYSA-L
IUPAC Name 4-oxo-2H,4H-benzo[d]1,3-dioxa-2-bismacyclohexan-2-yl hydrate
SMILES O.O=C1O[Bi]OC2=CC=CC=C12

Specifications

pH 2.7-5.0
CAS Max % 100%
Physical Form Crystalline Powder, Needles or Crystals
Quantity 25 g
Grade USP
Loss on Drying 1%
Packaging Poly Bottle
Chemical Name or Material Bismuth Subsalicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.