Learn More
MOLTOX Molecular Toxicology Inc Bismuth Sulfite Agar, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator/Dark
SDP
Catalog No. 50948807
Encompass_Preferred
Bismuth Sulfite Agar is a highly selective medium used for the isolation of Salmonella species, particularly Salmonella typhi in various types of samples including food, water and sewage. This product is not autoclaved in order to maintain its selective properties. This product is also light sensitive and must be protected from light. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-807 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S007
Bismuth Sulfite Agar is a highly selective medium used for the isolation of Salmonella species, particularly Salmonella typhi in various types of samples including food, water and sewage. This product is not autoclaved in order to maintain its selective properties. This product is also light sensitive and must be protected from light. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.