Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth titanium isopropoxide, 5% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog No. AA42840AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42840AC 25 mL
AA42840AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42840AC Supplier Thermo Scientific Chemicals Supplier No. 42840AC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338391-61-0
Molecular Formula C21H49BiO7Ti
MDL Number MFCD01865878
Synonym bismuthane titanium, bismuth-titanium 1/1, bismuth, compd. with titanium 1:1, bismuth titanium isopropoxide w/v in isopropanol, bismuth titanium isopropoxide w/v in isopropanol, packaged under argon in resealable chemseal bottles

Specifications

Color Yellow
Packaging ChemSeal™ bottles
Linear Formula BiTi[OCH(CH3)2]7
Quantity 25 mL
UN Number UN1219
Sensitivity Moisture Sensitive
Formula Weight 670.48
Physical Form Liquid
Chemical Name or Material Bismuth titanium isopropoxide
Hazard Category Danger
Hazard Statement GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
storageNote1
  • Packaged under Argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.