Learn More
Strem, An Ascensus Company CAS# 10035-06-0. 100G. Bismuth(III) nitrate pentahydrate (99.999%-Bi) PURATREM. MFCD00149157
SDP
Catalog No. 501963621
Encompass
CAS# 10035-06-0. 100G. Bismuth(III) nitrate pentahydrate (99.999%-Bi) PURATREM. MFCD00149157. Molecular Formula: Bi(NO3)3∙5H2O. Molecular Weight: 394.99 (485.07). Color/Form: white xtl. Stability: moisture sensitive. Strem# 93-8303. www.strem.com/catalog/v3/93-8303/
Catalog No. 50-196-3621 Supplier Strem, An Ascensus Company Supplier No. 938303100G
CAS# 10035-06-0. 100G. Bismuth(III) nitrate pentahydrate (99.999%-Bi) PURATREM. MFCD00149157. Molecular Formula: Bi(NO3)3∙5H2O. Molecular Weight: 394.99 (485.07). Color/Form: white xtl. Stability: moisture sensitive. Strem# 93-8303. www.strem.com/catalog/v3/93-8303/