Learn More
Medchemexpress LLC Bisoprolol-d5 | 1189881-87-5 | 1 MG
SDP
Catalog No. 5000340487
Encompass_Preferred
Bisoprolol-d5 is a deuterium-labeled variant of Bisoprolol, a potent, selective β1-adrenergic receptor blocker. It has potential for research in hypertension, coronary artery disease, and stable ventricular dysfunction. This compound is also used as a tracer and an internal standard for quantitative analysis. Deuteration can impact drug pharmacokinetic and metabolic profiles.
  • Deuterium labeled Bisoprolol
  • Potent, selective β1-adrenergic receptor blocker
  • Minimal β2-receptor activity
  • Used in research for hypertension, coronary artery disease, and stable ventricular dysfunction
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
Catalog No. 50-003-40487 Supplier Medchemexpress LLC Supplier No. HY129029S1MG
Bisoprolol-d5 is a deuterium-labeled variant of Bisoprolol, a potent, selective β1-adrenergic receptor blocker. It has potential for research in hypertension, coronary artery disease, and stable ventricular dysfunction. This compound is also used as a tracer and an internal standard for quantitative analysis. Deuteration can impact drug pharmacokinetic and metabolic profiles.
  • Deuterium labeled Bisoprolol
  • Potent, selective β1-adrenergic receptor blocker
  • Minimal β2-receptor activity
  • Used in research for hypertension, coronary artery disease, and stable ventricular dysfunction
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)