Learn More
Apexbio Technology LLC Bisoprolol fumarate 104344-23-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089390
Encompass_Preferred
Bisoprolol fumarate (CAS 104344-23-2) is a selective 1-adrenergic receptor antagonist that preferentially inhibits 1-adrenergic signaling with minimal activity at 2 receptors By suppressing 1 receptor-mediated pathways it reduces both heart rate and myocardial contractility following oral administration The compound is utilized in pharmacological and physiological studies to investigate mechanisms underlying hypertension coronary artery disease and stable ventricular dysfunction as well as to evaluate therapeutic interventions that target sympathetic nervous system modulation in cardiovascular research contexts
Catalog No. 50-000-89390 Supplier Apexbio Technology LLC Supplier No. B13545.1
Bisoprolol fumarate (CAS 104344-23-2) is a selective 1-adrenergic receptor antagonist that preferentially inhibits 1-adrenergic signaling with minimal activity at 2 receptors By suppressing 1 receptor-mediated pathways it reduces both heart rate and myocardial contractility following oral administration The compound is utilized in pharmacological and physiological studies to investigate mechanisms underlying hypertension coronary artery disease and stable ventricular dysfunction as well as to evaluate therapeutic interventions that target sympathetic nervous system modulation in cardiovascular research contexts