Learn More
Medchemexpress LLC Bisoprolol hemifumarate | 104344-23-2 | 99.8% | C18H31NO4.1/2C4H4O4 | 250 MG
SDP
Catalog No. 5000359044
Encompass_Preferred
Bisoprolol hemifumarate is a potent, selective, and orally active β1-adrenergic receptor blocker with minimal activity on β2-receptors. This compound is used in research for hypertension, coronary artery disease, and stable ventricular dysfunction studies.
  • Potent and selective β1-adrenergic receptor blocker
  • Orally active
  • Minimal β2-receptor activity
  • Protects myocardial cells from ischemia/reperfusion injury
  • Reduces ROS production and apoptosis
  • Increases AKT and GSK3β phosphorylation
  • Supports research in hypertension
  • Supports research in coronary artery disease
  • Supports research in stable ventricular dysfunction
  • Increases left ventricular ejection fraction
  • Decreases heart rate
  • Shows protective effects against cadmium-induced myocardial toxicity
  • Reverses small conductance calcium-activated potassium channel remodeling
Catalog No. 50-003-59044 Supplier Medchemexpress LLC Supplier No. 250MGHYB0076
Bisoprolol hemifumarate is a potent, selective, and orally active β1-adrenergic receptor blocker with minimal activity on β2-receptors. This compound is used in research for hypertension, coronary artery disease, and stable ventricular dysfunction studies.
  • Potent and selective β1-adrenergic receptor blocker
  • Orally active
  • Minimal β2-receptor activity
  • Protects myocardial cells from ischemia/reperfusion injury
  • Reduces ROS production and apoptosis
  • Increases AKT and GSK3β phosphorylation
  • Supports research in hypertension
  • Supports research in coronary artery disease
  • Supports research in stable ventricular dysfunction
  • Increases left ventricular ejection fraction
  • Decreases heart rate
  • Shows protective effects against cadmium-induced myocardial toxicity
  • Reverses small conductance calcium-activated potassium channel remodeling