Learn More
Cayman Chemical Bisoprolol (hemifumarate), Purity: ≥98%, CAS: 104344-23-2, 50 mg
SDP
Catalog No. 502994829
Encompass_Preferred
A β1-AR antagonist (Ki = 25 nM for the human receptor); selective for β1- over β2-ARs (Ki = 480 nM for the human receptor in a radioligand binding assay); binds to rat ventricular myocytes and heart membranes that endogenously express β1-ARs and β1- and β2-ARs, respectively (Kis = 20 and 38.1 nM, respectively); inhibits isoproterenol-induced increases in heart rate and myocardial contractility in conscious dogs at 0.3 mg/kg; decreases blood pressure and heart rate in SHRs at 7.5 mg/kg twice per day over 19 weeks; inhibits SARS-CoV-2 Mpro (IC50 = 118.5 µg/ml) and reduces viral infectivity in SARS-CoV-2 infected Vero E6 cells (IC50 = 15.917 µg/ml)
Catalog No. 50-299-4829 Supplier Cayman Chemical Supplier No. 2382750 MG
A β1-AR antagonist (Ki = 25 nM for the human receptor); selective for β1- over β2-ARs (Ki = 480 nM for the human receptor in a radioligand binding assay); binds to rat ventricular myocytes and heart membranes that endogenously express β1-ARs and β1- and β2-ARs, respectively (Kis = 20 and 38.1 nM, respectively); inhibits isoproterenol-induced increases in heart rate and myocardial contractility in conscious dogs at 0.3 mg/kg; decreases blood pressure and heart rate in SHRs at 7.5 mg/kg twice per day over 19 weeks; inhibits SARS-CoV-2 Mpro (IC50 = 118.5 µg/ml) and reduces viral infectivity in SARS-CoV-2 infected Vero E6 cells (IC50 = 15.917 µg/ml)