Learn More
Selleck Chemical LLC Bisphenol A
SDP
Supplier:  Selleck Chemical LLC S60071G
Encompass_Preferred
Bisphenol A is a starting material for the synthesis of plastics primarily certain polycarbonates and epoxy resins as well as some polysulfones and certain niche materials It exhibits estrogen-mimicking hormone-like properties
Catalog No. 50-000-03307