Learn More
Medchemexpress LLC Bisphenol A bis(diphenyl phosphate) | 5945-33-5 | 692.63 g·mol⁻¹ | C39H34O8P2 | 25 MG
SDP
Catalog No. 5000283653
Encompass_Preferred
Bisphenol A bis(diphenyl phosphate) is an organophosphate flame retardant used to reduce flammability in polymers and other materials. It exhibits both condensed-phase and gas-phase flame-retardant actions and is supplied as a research sample; identifiers are provided for traceability (CAS 5945-33-5; C39H34O8P2; MW 692.63 g·mol⁻¹).
  • Acts in condensed and gas phases.
  • Designed for flame-retardant polymer formulations.
  • Supplied in small sample quantities for research.
  • Provides clear chemical identifiers for traceability.
  • Purity and storage information available on the certificate of analysis.
Catalog No. 50-002-83653 Supplier Medchemexpress LLC Supplier No. HYW12956125MG
Bisphenol A bis(diphenyl phosphate) is an organophosphate flame retardant used to reduce flammability in polymers and other materials. It exhibits both condensed-phase and gas-phase flame-retardant actions and is supplied as a research sample; identifiers are provided for traceability (CAS 5945-33-5; C39H34O8P2; MW 692.63 g·mol⁻¹).
  • Acts in condensed and gas phases.
  • Designed for flame-retardant polymer formulations.
  • Supplied in small sample quantities for research.
  • Provides clear chemical identifiers for traceability.
  • Purity and storage information available on the certificate of analysis.