Learn More
Medchemexpress LLC Bisphenol A bis(diphenyl phosphate) | 5945-33-5 | MFCD09753077 | 692.63 g/mol | C39H34O8P2 | 50 MG
SDP
Catalog No. 5000282957
Encompass_Preferred
Bisphenol A bis(diphenyl phosphate) is an organophosphorus flame retardant used to impart flame resistance to polymers and materials. It demonstrates a significant condensed-phase action in addition to gas-phase flame inhibition, and is provided for research, formulation, and material testing.
  • Acts in both condensed phase and gas phase.
  • Suitable for polymer and coating formulations.
  • Available in multiple small sizes for laboratory use.
  • Datasheet and certificate of analysis available for storage and handling guidance.
  • Useful for flame retardancy evaluation and material testing.
Catalog No. 50-002-82957 Supplier Medchemexpress LLC Supplier No. HYW12956150MG
Bisphenol A bis(diphenyl phosphate) is an organophosphorus flame retardant used to impart flame resistance to polymers and materials. It demonstrates a significant condensed-phase action in addition to gas-phase flame inhibition, and is provided for research, formulation, and material testing.
  • Acts in both condensed phase and gas phase.
  • Suitable for polymer and coating formulations.
  • Available in multiple small sizes for laboratory use.
  • Datasheet and certificate of analysis available for storage and handling guidance.
  • Useful for flame retardancy evaluation and material testing.