Learn More
Medchemexpress LLC Bisphenol A bis(diphenyl phosphate) | 5945-33-5 | MFCD09753077 | >97.0% | 692.63 g·mol-1 | C39H34O8P2 | 250 MG
SDP
Catalog No. 5000282619
Encompass_Preferred
Bisphenol A bis(diphenyl phosphate) is an organophosphate flame retardant provided as a research-grade reagent. It is used to impart flame resistance to polymers and materials and displays both condensed-phase and gas-phase flame-retardant actions. Typical identifiers include CAS 5945-33-5, formula C39H34O8P2, and molecular weight 692.63 g·mol-1.
Catalog No. 50-002-82619 Supplier Medchemexpress LLC Supplier No. HYW129561250MG
Add to Cart
Add to Cart
Bisphenol A bis(diphenyl phosphate) is an organophosphate flame retardant provided as a research-grade reagent. It is used to impart flame resistance to polymers and materials and displays both condensed-phase and gas-phase flame-retardant actions. Typical identifiers include CAS 5945-33-5, formula C39H34O8P2, and molecular weight 692.63 g·mol-1.