Learn More
Medchemexpress LLC Bisphenol A-d14 | 120155-79-5 | 99.7% | 242.37 | 1 MG
SDP
Catalog No. 5000387673
Encompass_Preferred
Bisphenol A-d14 is a deuterium-labeled Bisphenol A. Bisphenol A is a phenolic, organic synthetic compound widely used in the production of polycarbonate plastics and epoxy resins. It is classified as an endocrine-disrupting compound (EDC) and is associated with various diseases, including cardiovascular diseases, respiratory diseases, diabetes, kidney diseases, obesity, and reproductive disorders.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-87673 Supplier Medchemexpress LLC Supplier No. HY18260S61MG
Bisphenol A-d14 is a deuterium-labeled Bisphenol A. Bisphenol A is a phenolic, organic synthetic compound widely used in the production of polycarbonate plastics and epoxy resins. It is classified as an endocrine-disrupting compound (EDC) and is associated with various diseases, including cardiovascular diseases, respiratory diseases, diabetes, kidney diseases, obesity, and reproductive disorders.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.