Learn More
Medchemexpress LLC Bisphenol A-d16 | 96210-87-6 | 99.9% | 244.38 | 100 MG
SDP
Catalog No. 5000387622
Encompass_Preferred
Bisphenol A-d16 is the deuterium labeled Bisphenol A, a phenolic, organic synthetic compound widely used in the production of polycarbonate plastics and epoxy resins. It is often classified as an endocrine-disrupting compound (EDC) and is associated with various diseases, including cardiovascular diseases, respiratory diseases, diabetes, kidney diseases, obesity, and reproductive disorders.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-87622 Supplier Medchemexpress LLC Supplier No. HY18260S100MG
Bisphenol A-d16 is the deuterium labeled Bisphenol A, a phenolic, organic synthetic compound widely used in the production of polycarbonate plastics and epoxy resins. It is often classified as an endocrine-disrupting compound (EDC) and is associated with various diseases, including cardiovascular diseases, respiratory diseases, diabetes, kidney diseases, obesity, and reproductive disorders.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS