Learn More
Medchemexpress LLC Bisphenol A-d16 | 96210-87-6 | MFCD29073707 | 100.0% | 244.38 g/mol | C15D16O2 | 10 MG
SDP
Catalog No. 5000294838
Encompass_Preferred
Bisphenol A-d16 is a deuterium-labeled isotopologue of bisphenol A intended for research and analytical applications. It is used as a stable isotope internal standard and tracer in quantitative mass spectrometry and toxicology studies, providing consistent, traceable results for analytical workflows.
  • Deuterium-labeled internal standard for quantitative assays.
  • High isotopic enrichment for accurate mass spectrometry measurements.
  • High chemical purity suitable for analytical use.
  • Supplied in a small vial for trace-level standard preparation.
  • Useful for isotope dilution and tracer studies.
Catalog No. 50-002-94838 Supplier Medchemexpress LLC Supplier No. HY18260S10MG
Bisphenol A-d16 is a deuterium-labeled isotopologue of bisphenol A intended for research and analytical applications. It is used as a stable isotope internal standard and tracer in quantitative mass spectrometry and toxicology studies, providing consistent, traceable results for analytical workflows.
  • Deuterium-labeled internal standard for quantitative assays.
  • High isotopic enrichment for accurate mass spectrometry measurements.
  • High chemical purity suitable for analytical use.
  • Supplied in a small vial for trace-level standard preparation.
  • Useful for isotope dilution and tracer studies.