Learn More
Medchemexpress LLC Bisphenol A-d4 | 102438-62-0 | 10 MG
SDP
Catalog No. 5000294687
Encompass_Preferred
Bisphenol A-d4 is a deuterium-labeled analog of bisphenol A supplied as a solid for use as an internal standard and tracer in analytical and research workflows. It is intended for quantitative analysis and method validation using NMR, GC-MS, and LC-MS, and is provided in a 10 mg package.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Provided as a white to off-white solid in 10 mg packaging.
  • Molecular weight 232.31 g/mol; CAS 102438-62-0.
  • Store powder at -20°C for long-term stability.
Catalog No. 50-002-94687 Supplier Medchemexpress LLC Supplier No. HY18260S410MG
Bisphenol A-d4 is a deuterium-labeled analog of bisphenol A supplied as a solid for use as an internal standard and tracer in analytical and research workflows. It is intended for quantitative analysis and method validation using NMR, GC-MS, and LC-MS, and is provided in a 10 mg package.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Provided as a white to off-white solid in 10 mg packaging.
  • Molecular weight 232.31 g/mol; CAS 102438-62-0.
  • Store powder at -20°C for long-term stability.