Learn More
Medchemexpress LLC Bisphenol A-d4 | 102438-62-0 | 232.31 | 5 MG
SDP
Catalog No. 5000387603
Encompass_Preferred
Bisphenol A-d4 is the deuterium-labeled Bisphenol A. Bisphenol A is a phenolic, organic synthetic compound extensively used in the production of polycarbonate plastics and epoxy resins. It is classified as a reproductive, developmental, and systemic toxicant, often considered an endocrine-disrupting compound (EDC). Bisphenol A has been linked to numerous diseases, including cardiovascular diseases, respiratory diseases, diabetes, kidney diseases, obesity, and reproductive disorders.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-87603 Supplier Medchemexpress LLC Supplier No. HY18260S45MG
Bisphenol A-d4 is the deuterium-labeled Bisphenol A. Bisphenol A is a phenolic, organic synthetic compound extensively used in the production of polycarbonate plastics and epoxy resins. It is classified as a reproductive, developmental, and systemic toxicant, often considered an endocrine-disrupting compound (EDC). Bisphenol A has been linked to numerous diseases, including cardiovascular diseases, respiratory diseases, diabetes, kidney diseases, obesity, and reproductive disorders.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.