Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bisphenol A-d6 | 86588-58-1 | MFCD00224249 | 99.0% | 234.32 g/mol | C15H10D6O2 | 10 MG
SDP

Catalog No. 5000295453
Encompass_Preferred

Bisphenol A-d6 is a deuterium-labeled analogue of bisphenol A supplied as a solid analytical standard for use as an internal standard in mass spectrometry and isotopic tracing studies. It is intended for analytical workflows requiring a stable isotope-labeled reference and small laboratory pack sizes.

  • Deuterium-labeled internal standard for mass spectrometry.
  • Molecular formula C15H10D6O2 and molecular weight 234.32 g/mol.
  • High chemical purity (99.0%).
  • Available in small pack sizes such as 10 MG for analytical use.
  • Recommended storage: powder at -20°C for long-term stability; in solvent at -80°C for extended storage.

Catalog No. 50-002-95453 Supplier Medchemexpress LLC Supplier No. HY18260S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bisphenol A-d6 is a deuterium-labeled analogue of bisphenol A supplied as a solid analytical standard for use as an internal standard in mass spectrometry and isotopic tracing studies. It is intended for analytical workflows requiring a stable isotope-labeled reference and small laboratory pack sizes.

  • Deuterium-labeled internal standard for mass spectrometry.
  • Molecular formula C15H10D6O2 and molecular weight 234.32 g/mol.
  • High chemical purity (99.0%).
  • Available in small pack sizes such as 10 MG for analytical use.
  • Recommended storage: powder at -20°C for long-term stability; in solvent at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.