Learn More
Medchemexpress LLC Bisphenol A-d8 | 92739-58-7 | 99.3% | 236.34 | 1 MG
SDP
Catalog No. 5000387686
Encompass_Preferred
Bisphenol A-d8 is the deuterium labeled Bisphenol A. Bisphenol A is a phenolic, organic synthetic compound widely used in the production of polycarbonate plastics and epoxy resins. It is classified as a reproductive, developmental, and systemic toxicant, often considered an endocrine-disrupting compound (EDC). Bisphenol A is linked to various health issues including cardiovascular diseases, respiratory diseases, diabetes, kidney diseases, obesity, and reproductive disorders.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-87686 Supplier Medchemexpress LLC Supplier No. HY18260S31MG
Bisphenol A-d8 is the deuterium labeled Bisphenol A. Bisphenol A is a phenolic, organic synthetic compound widely used in the production of polycarbonate plastics and epoxy resins. It is classified as a reproductive, developmental, and systemic toxicant, often considered an endocrine-disrupting compound (EDC). Bisphenol A is linked to various health issues including cardiovascular diseases, respiratory diseases, diabetes, kidney diseases, obesity, and reproductive disorders.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs.