Learn More
Medchemexpress LLC Bisphenol A-d8 | 92739-58-7 | 99.3% | 236.34 | 5 MG
SDP
Catalog No. 5000387552
Encompass_Preferred
Bisphenol A-d8 is a deuterium-labeled version of Bisphenol A. Bisphenol A is a phenolic, organic synthetic compound extensively used in the production of polycarbonate plastics and epoxy resins. It is classified as an endocrine-disrupting compound (EDC) and is associated with various health issues, including cardiovascular diseases, respiratory diseases, diabetes, kidney diseases, obesity, and reproductive disorders.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis using methods like NMR, GC-MS, or LC-MS.
Catalog No. 50-003-87552 Supplier Medchemexpress LLC Supplier No. HY18260S35MG
Bisphenol A-d8 is a deuterium-labeled version of Bisphenol A. Bisphenol A is a phenolic, organic synthetic compound extensively used in the production of polycarbonate plastics and epoxy resins. It is classified as an endocrine-disrupting compound (EDC) and is associated with various health issues, including cardiovascular diseases, respiratory diseases, diabetes, kidney diseases, obesity, and reproductive disorders.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis using methods like NMR, GC-MS, or LC-MS.