Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bisphenol A ethoxylate diacrylate

Catalog No. 413550250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-355-0250ML 250 mL
41-355-01L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-355-0250ML Supplier Sigma Aldrich Supplier No. 413550250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bisphenol A ethoxylate diacrylate is a cross-linking monomer that is used to improve the mechanical properties of the polymeric system. It is majorly used in the formation of solid polymer based electrolytes which can be used for electrochemical applications.

Specifications

CAS 64401-02-1
Density 1.146 g/mL (at 25°C)
Refractive Index n20/D 1.545
Linear Formula [H2C=CHCO2(CH2CH2O)nC6H4-4-]2 C(CH3)2
MDL Number MFCD03793133
Quantity 250 mL
Molecular Weight (g/mol) 468
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.