Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bisphenol A glycerolate (1 glycerol/phenol) diacrylate

Catalog No. 411167250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-116-7250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-116-7250ML Supplier Sigma Aldrich Supplier No. 411167250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

UV curable cross-linking agent. Component of curable inks, coatings, varnishes and lacquers.

Specifications

CAS 4687-94-9
Density 1.18 g/mL (at 25°C)
Molecular Formula C27H32O8
Refractive Index n20/D 1.557 (literature)
Linear Formula C27H32O8
Quantity 250 mL
MDL Number MFCD01317012
Synonym 2,2-Bis[4-(3-acryloyloxy-2-hydroxypropoxy)phenyl]propane; 2,2-Bis[acryloyloxy(2 -hydroxypropyloxy)phenyl]propane; Bisphenol A glycerolate diacrylate
Molecular Weight (g/mol) 484.54
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.