Learn More
Medchemexpress LLC Bisphenol A monosulfate-d6 sodium | 2733721-65-6 | C15H9D6NaO5S | 1 MG
SDP
Catalog No. 5000353581
Encompass_Preferred
Bisphenol A monosulfate-d6 (sodium) is the deuterium-labeled Bisphenol A monosulfate sodium, intended for research use only. This compound serves as a tracer and an internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS. Deuterium substitution is recognized for its potential influence on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis.
  • Stable heavy isotopes for quantitation during drug development.
  • Deuteration can potentially affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-53581 Supplier Medchemexpress LLC Supplier No. HY142555S1MG
Bisphenol A monosulfate-d6 (sodium) is the deuterium-labeled Bisphenol A monosulfate sodium, intended for research use only. This compound serves as a tracer and an internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS. Deuterium substitution is recognized for its potential influence on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis.
  • Stable heavy isotopes for quantitation during drug development.
  • Deuteration can potentially affect pharmacokinetic and metabolic profiles of drugs.