Learn More
Medchemexpress LLC Bisphenol AP-d5 | 2469555-57-3 | 95.0% | 295.39 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW008628S1MG
Encompass_Preferred
Bisphenol AP-d5 is a deuterium-labeled form of Bisphenol AP. This compound is intended for research use only and is not sold to patients. Deuteration, the incorporation of stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, is increasingly recognized for its potential to influence the pharmacokinetic and metabolic profiles of drugs. Such labeled compounds are largely used as tracers for quantitative analysis during the drug development process.
  • Application as a tracer
  • Internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS
  • Deuterium substitution can impact the pharmacokinetics of pharmaceuticals
Catalog No. 50-003-66671