Learn More
Medchemexpress LLC Bisphenol C 100g
SDP
Catalog No. 5000206635
Encompass_Preferred
Bisphenol C is an estrogen receptor- (ER ) agonist and an ER antagonist with IC50 values of 2 65 nM for ER and 1 94 nM for ER Bisphenol C is a material of manufacturing polyester polymers like polycarbonate is widely used in daily items like water bottles food packaging textile and so on[1][2][3]
Catalog No. 50-002-06635 Supplier Medchemexpress LLC Supplier No. HYW114419100G
Bisphenol C is an estrogen receptor- (ER ) agonist and an ER antagonist with IC50 values of 2 65 nM for ER and 1 94 nM for ER Bisphenol C is a material of manufacturing polyester polymers like polycarbonate is widely used in daily items like water bottles food packaging textile and so on[1][2][3]