Learn More
Medchemexpress LLC Bisphenol F | 620-92-8 | MFCD00002385 | 99.9% | 200.24 g/mol | C13H12O2 | 1 ML
SDP
Catalog No. 5000302516
Encompass_Preferred
Bisphenol F (4,4'-dihydroxydiphenylmethane, CAS 620-92-8) is a bisphenol derivative used in research on endocrine disruption and oxidative stress pathways. Offered as a high-purity solid and as a ready-to-use 10 mM solution in DMSO (1 mL), it is suitable for biochemical assays, cell studies, and analytical methods.
  • High purity (99.93%) suitable for analytical and research applications.
  • Available as a 10 mM solution in DMSO for immediate use.
  • Also available as solid in multiple gram-scale quantities for method development.
  • Molecular weight 200.24 g/mol and formula C13H12O2 for precise calculations.
  • Useful for studying reactive oxygen species and AKT/GSK3β signaling pathways.
Catalog No. 50-003-02516 Supplier Medchemexpress LLC Supplier No. HYW01490110MM/1ML
Bisphenol F (4,4'-dihydroxydiphenylmethane, CAS 620-92-8) is a bisphenol derivative used in research on endocrine disruption and oxidative stress pathways. Offered as a high-purity solid and as a ready-to-use 10 mM solution in DMSO (1 mL), it is suitable for biochemical assays, cell studies, and analytical methods.
  • High purity (99.93%) suitable for analytical and research applications.
  • Available as a 10 mM solution in DMSO for immediate use.
  • Also available as solid in multiple gram-scale quantities for method development.
  • Molecular weight 200.24 g/mol and formula C13H12O2 for precise calculations.
  • Useful for studying reactive oxygen species and AKT/GSK3β signaling pathways.