Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bisphenol F acetate propionate, epoxy curative, EC-298

Catalog No. p-7080194
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4672414 25 g
AA4672422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4672414 Supplier Thermo Scientific Chemicals Supplier No. 04672414
Only null left
Add to Cart

Well suited for use in epoxy/(meth)acrylate hybrid thermoset compositions | CAS: 1071466-53-9 | C18H18O4 | 298.338 g/mol

Bisphenol F acetate propionate does not impede free radical cure, thermoset adhesives, used as curative for epoxy/(meth) acrylate hybrids, hybrid epoxy/free radical thermosets. Bisphenol F is used to make epoxy resins and coatings for various applications, such as lacquers, varnishes, liners, adhesives, plastics, water pipes, dental sealants, and food packaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071466-53-9
Molecular Formula C18H18O4
Molecular Weight (g/mol) 298.338
MDL Number MFCD22380705
InChI Key JARUUDOWTQBKKU-UHFFFAOYSA-N
Synonym 4-4-acetyloxy phenyl methyl phenyl propanoate, bisphenol f acetate propionate, epoxy curative, ec-298
PubChem CID 59317436
IUPAC Name [4-[(4-acetyloxyphenyl)methyl]phenyl] propanoate
SMILES CCC(=O)OC1=CC=C(C=C1)CC2=CC=C(C=C2)OC(=O)C

Specifications

Color Yellow
Physical Form Pale yellow liquid
Density 1.13
Solubility Information Soluble in ethanol,ether,chloroform,alkali; slightly soluble in DMSO; insoluble in carbon disulfide.
Quantity 25 g
Formula Weight 298.34
Flash Point 150°C (302°F)
Chemical Name or Material Bisphenol F acetate propionate, Epoxy curative, EC-298
Hazard Category H302-H319
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.