Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BisphenolA (2,3-dihydroxypropyl) glycidyl ether, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015445 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5445 25 mg
11-101-5444 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5445 Supplier MilliporeSigma Supelco Supplier No. 7341725MG
Add to Cart
Add to Cart

Bisphenol A (2,3-dihydroxypropyl) glycidyl ether can be obtained as a product of the reaction of bisphenol A with epichlorohydrin.It is generally used as a substrate to prepare epoxy resins, which is used in coatings and adhesives.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 76002-91-0
Physical Form Neat
Linear Formula C21H26O5
MDL Number MFCD03453592
Synonym 2-[4-(2,3-Dihydroxypropyloxy)phenyl]-2-[4-(glycidyloxy)phenyl]propane; BADGE-H2O
Type Analytical Standard
Quantity 25 mg
Formula Weight 358.43
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.