Learn More
Sielc Technologies Inc BIST A+ COLUMN 4.6X150MM 5UM
Catalog No. NC2170924
Encompass
BIST A+ Column 4.6x150mm 5um 100A
This item is not returnable. View return policy
Catalog No. NC2170924 Supplier Sielc Technologies Inc Supplier No. TAP46.150.0510
BIST A+ Column 4.6x150mm 5um 100A