Learn More
Strem, An Ascensus Company CAS# 19999-87-2. 1g. Bis(tricyclohexylphosphine)nickel(II) chloride, 99%. MFCD11973802
SDP
Catalog No. 50245454
Encompass
CAS# 19999-87-2. 1g. Bis(tricyclohexylphosphine)nickel(II) chloride, 99%. MFCD11973802. Molecular Weight: 690.46. Molecular Formula: [(C6H11)3P]2NiCl2. Color/form: dark red-purple xtls. Strem# 28-0091. http://www.strem.com/catalog/v/28-0091/
Catalog No. 50-245-454 Supplier Strem, An Ascensus Company Supplier No. 2800911G
CAS# 19999-87-2. 1g. Bis(tricyclohexylphosphine)nickel(II) chloride, 99%. MFCD11973802. Molecular Weight: 690.46. Molecular Formula: [(C6H11)3P]2NiCl2. Color/form: dark red-purple xtls. Strem# 28-0091. http://www.strem.com/catalog/v/28-0091/