Learn More
AdipoGen Bistrifluron
SDP
Supplier:  AdipoGen CDXB0125M050
Encompass_Preferred
Chemical. CAS 201593-84-2. Formula C16H7ClF8N2O2. MW 446.7. Synthetic. Insecticide. Insect growth regulator. Inhibitor of chitin biosynthesis, type 0. Compound can be used as analytical reference material.
Catalog No. 50-256-1667