Learn More
Cayman Chemical Bisubstrate Inhibitor 78, Purity: ≥98%, CAS: 2368247-39-4, 1 mg
SDP
Catalog No. 502940745
Encompass_Preferred
An NNMT inhibitor (IC50 = 1.41 µM); binds the NNMT active site in the binding pockets for SAM and nicotinamide; selective for NNMT over NSD2 and PRMT1 (IC50s = >50 µM for both); reduces MNA levels in, and inhibits proliferation of, HSC-2 oral cancer cells at 100 µM
Catalog No. 50-294-0745 Supplier Cayman Chemical Supplier No. 285671 MG
An NNMT inhibitor (IC50 = 1.41 µM); binds the NNMT active site in the binding pockets for SAM and nicotinamide; selective for NNMT over NSD2 and PRMT1 (IC50s = >50 µM for both); reduces MNA levels in, and inhibits proliferation of, HSC-2 oral cancer cells at 100 µM