Learn More
Cayman Chemical BIsubstrate InhIbItr 78 500ug
SDP
Catalog No. 502938518
Encompass_Preferred
An NNMT inhibitor (IC50 = 1.41 µM); binds the NNMT active site in the binding pockets for SAM and nicotinamide; selective for NNMT over NSD2 and PRMT1 (IC50s = >50 µM for both); reduces MNA levels in, and inhibits proliferation of, HSC-2 oral cancer cells at 100 µM
Catalog No. 50-293-8518 Supplier Cayman Chemical Supplier No. 28567500 UG
An NNMT inhibitor (IC50 = 1.41 µM); binds the NNMT active site in the binding pockets for SAM and nicotinamide; selective for NNMT over NSD2 and PRMT1 (IC50s = >50 µM for both); reduces MNA levels in, and inhibits proliferation of, HSC-2 oral cancer cells at 100 µM