Learn More
STEMCELL Technologies BIT 9500 Serum Substitute, Size: 100 mL
SDP
Catalog No. NC9952276
Encompass
BIT 9500 Serum Substitute has been developed for use in applications where a serum-free culture medium of defined composition is required. This product contains bovine serum albumin (BSA), insulin, and transferrin (BIT) in Iscove’s MDM. It contains pre-screened batches of BSA that have been selected to support the optimal growth of human hematopoietic progenitor cells in serum-free media formulations. It is also suitable for the culture of mouse hematopoietic progenitor cells in serum-free conditions.
Catalog No. NC9952276 Supplier STEMCELL Technologies Supplier No. 09500
BIT 9500 Serum Substitute has been developed for use in applications where a serum-free culture medium of defined composition is required. This product contains bovine serum albumin (BSA), insulin, and transferrin (BIT) in Iscove’s MDM. It contains pre-screened batches of BSA that have been selected to support the optimal growth of human hematopoietic progenitor cells in serum-free media formulations. It is also suitable for the culture of mouse hematopoietic progenitor cells in serum-free conditions.