Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bitertanol | 55179-31-2 | MFCD00055508 | 98.7% | 337.42 g/mol | C20H23N3O2 | 250 MG
SDP

Catalog No. 5000228711
Encompass_Preferred

Bitertanol is a triazole-class antifungal research compound (CAS 55179-31-2) used in studies of antifungal activity and cytochrome P450 modulation. It has been reported as a CYP1A1, CYP2B, and CYP3A inducer in vivo and as a CYP1A inhibitor in vitro.

  • Triazole antifungal research compound.
  • Purity 98.7% (as supplied).
  • Molecular formula C20H23N3O2; molecular weight 337.42 g/mol.
  • Available in small research pack sizes, including 250 MG.
  • Suitable for studies of antifungal activity and P450 enzyme modulation.

Catalog No. 50-002-28711 Supplier Medchemexpress LLC Supplier No. HY118216250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bitertanol is a triazole-class antifungal research compound (CAS 55179-31-2) used in studies of antifungal activity and cytochrome P450 modulation. It has been reported as a CYP1A1, CYP2B, and CYP3A inducer in vivo and as a CYP1A inhibitor in vitro.

  • Triazole antifungal research compound.
  • Purity 98.7% (as supplied).
  • Molecular formula C20H23N3O2; molecular weight 337.42 g/mol.
  • Available in small research pack sizes, including 250 MG.
  • Suitable for studies of antifungal activity and P450 enzyme modulation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.