Learn More
Medchemexpress LLC HY-10809 5mg Medchemexpress, Bitopertin CAS:845614-11-1 Purity:>98%
SDP
Catalog No. 501968507
Encompass_Preferred
Medchemexpress, HY-10809 5mg Bitopertin CAS:845614-11-1 Bitopertin is a potent, noncompetitive glycine reuptake inhibitor, inhibits glycine uptake at human GlyT1 with a concentration exhibiting IC50 of 25 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8507 Supplier Medchemexpress LLC Supplier No. HY108095MG
Medchemexpress, HY-10809 5mg Bitopertin CAS:845614-11-1 Bitopertin is a potent, noncompetitive glycine reuptake inhibitor, inhibits glycine uptake at human GlyT1 with a concentration exhibiting IC50 of 25 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.