Learn More
Specifications
Specifications
| Chemical Name or Material | Bitterness Suppressor Flavor |
| Packaging | Amber Glass Bottle |
| Refractive Index | 1.342-1.442 |
| Quantity | 500 mL |
| Grade | Reagent |
| Physical Form | Viscous Liquid or Low Melting Solid |
CAUTION: For professional use only. Not for household use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
