Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Biuret
SCI_ED

Catalog No. S25199 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25199 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25199

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 108-19-0
Molecular Formula C2H5N3O2
Molecular Weight (g/mol) 103.081
InChI Key OHJMTUPIZMNBFR-UHFFFAOYSA-N
Synonym biuret, allophanamide, imidodicarbonic diamide, carbamylurea, ureidoformamide, dicarbonimidic diamide, allophanic acid amide, allophanimidic acid, dicarbamylamine, isobiuret
PubChem CID 7913
ChEBI CHEBI:18138
IUPAC Name carbamoylurea
SMILES C(=O)(N)NC(=O)N

Specifications

Assay Percent Range 97% w/v
Linear Formula NH2CONHCONH2
Density 1.467g/mL
Formula Weight 103.08
Packaging Solid
Quantity 50 g
Physical Form Solid
Chemical Name or Material Biuret
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.