Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biuret 99.0+%, TCI America™
SDP

Catalog No. B32281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B32281G Supplier TCI America Supplier No. B32281G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-19-0
Molecular Formula C2H5N3O2
Molecular Weight (g/mol) 103.081
MDL Number MFCD00007946
InChI Key OHJMTUPIZMNBFR-UHFFFAOYSA-N
Synonym biuret, allophanamide, imidodicarbonic diamide, carbamylurea, ureidoformamide, dicarbonimidic diamide, allophanic acid amide, allophanimidic acid, dicarbamylamine, isobiuret
PubChem CID 7913
ChEBI CHEBI:18138
IUPAC Name carbamoylurea
SMILES C(=O)(N)NC(=O)N

Specifications

Melting Point 193°C
Color White
Quantity 1 g
Formula Weight 103.08
Percent Purity ≥99.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Biuret [for Fertilizer Analysis]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.